Would you like to log in again?
Econo™ Littauer Scissors, Floor Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ECO™
21-358 21-363
89023-172PK 81.91 USD InStock
89023-172 89023-176
Econo™ Littauer Scissors, Floor Grade, Sklar
Scissors
Econo™ Littauer Suture scissors are a disposable, floor-grade instrument used for suture removal
Littauer Suture scissors have larger, less delicate tips than Spencer Stitch scissors, and are often used for removing larger sutures.
They should not be used for cutting tissue.
These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture, so it does not slip off the blade of the scissors prior to cutting.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty