Would you like to log in again?
Econo™ Rankin-Crile Hemostatic Forceps, Floor Grade, Sklar
Supplier: Sklar
Econo™
21-458 21-463
89023-208PK 147.18 USD InStock
89023-208 89023-210
Econo™ Rankin-Crile Hemostatic Forceps, Floor Grade, Sklar
Forceps
Econo™ Rankin-Crile Hemostatic Forceps are a standard clamp regularly used to hold tissue, or lumen.
Designed to stop bleeding, hemostatic clamps are differentiated by weight, size, and serration pattern. Econo™ Rankin-Crile Hemostatic Clamps feature a jaw that is 1/3 the length of the shank, similar to the Rankin Hemostat, but have serrations the entire length of the jaw, as seen in the Crile Hemostat.
Econo™ instruments are disposable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty