Would you like to log in again?
Econo™ Rochester-Pean Forceps, Floor Grade, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Econo™
21-495 21-477 21-471 21-470 21-473 21-485 21-481 21-487 21-492 21-498
89023-218PK 99.39 USD InStock
89023-218 89023-236 89023-226 89023-216 89023-232 89023-222 76192-556 89023-224 89023-220 76192-542
Econo™ Rochester-Pean Forceps, Floor Grade, Sklar®
Forceps
Econo™ Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The full horizontal serrations and availability in multiple lengths make these forceps a versatile instrument used in multiple procedures. They are available curved or straight. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty