Would you like to log in again?
Econo™ Foerster Sponge Forceps, Floor Grade, Sklar
Supplier: Sklar
Econo™
21-586 21-577
89023-244PK 253.27 USD InStock
89023-244 89023-246
Econo™ Foerster Sponge Forceps, Floor Grade, Sklar
Forceps
Econo™ Foerster Sponge Forceps, which may be known as Foerster-Ballenger Sponge Forceps, are used in surgical procedures to hold gauze squares that will absorb excess fluids and blood from the surgical site and hemostasis.
The ratcheted locking mechanism of these finger ring forceps grip the gauze and hold it in place. Foerster Sponge Forceps that are longer in length are often used in bariatric surgeries and the curved version may be used for removing the placenta in OB/GYN procedures.
Econo™ instruments are disposable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty