Would you like to log in again?
Econo™ Adson-Brown Tissue Forceps, Floor Grade, Sklar
Supplier: Sklar
Econo™
21-778
89023-294PK 128.52 USD InStock
89023-294
Econo™ Adson-Brown Tissue Forceps, Floor Grade, Sklar
Forceps
Econo™ Adson-Brown Tissue Forceps are designed for holding and manipulating delicate tissues
These forceps have a wide, flat thumb grasp area. The jaws are short and the tips are narrow. They are similar to standard Adson forceps, however, Adson-Brown forceps feature multiple rows of teeth, 7 x 7 or 9 x 9.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty