Would you like to log in again?
TC Derf Needle Holder, OR Grade, Sklar
Supplier: Sklar
21-8004
89023-312EA 293.79 USD InStock
89023-312
TC Derf Needle Holder, OR Grade, Sklar
Forceps
Sklar's® TC Derf Needle Holder features short jaws and delicate serrations that hold small needles, as well as 5-0 and 6-0 sutures.It is used in ophthalmic, dental, or plastic surgery procedures, the shorter length allows the Derf Needle Holder to be easily controlled in smaller surgical areas.
  • Stay sharp for a longer period of time
  • Tungsten carbide inserts provides maximizes control and precision
Censitrac Ready products are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty