Would you like to log in again?
TC Olsen-Hegar Needle Holder, OR Grade, Sklar
Supplier: Sklar
21-8029 21-8025 21-8026 21-8027 21-8028
89023-346EA 370.81 USD InStock
89023-346 89023-348 89023-354 89023-350 89023-352
TC Olsen-Hegar Needle Holder, OR Grade, Sklar
Forceps
Olsen-Hegar TC Needle Holder is a ratcheted, finger ring instrument with built in scissors
  • Censitrac Ready products are industry standard and constructed of stainless steel
  • Latex-free
Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.
It is used with intermediate weight sutures and in plastic surgery procedures.
Delivery information: Ordering Information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty