Would you like to log in again?
TC Castroviejo Needle Holder, OR Grade, Sklar®
Supplier: Sklar
21-8046 21-8047 21-8043 21-8040 21-8042 21-8045 21-8048 21-8044
76192-660EA 709.67 USD InStock
76192-660 89023-380 89023-376 89023-378 89023-382 89023-384 76192-662 89023-374
TC Castroviejo Needle Holder, OR Grade, Sklar®
Forceps
The Sklar® Tungsten Carbide Castroviejo Needle Holder is a multipurpose microsurgical instrument that is often used in ophthalmic and plastic surgeries.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The Castroviejo Needle Holder features a double spring, flat handle, and it is available with straight or curved jaws. It is also available with or without a locking mechanism. The small tips of this needle holder are ideal for handling small, delicate needles and sutures. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar® permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty