Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
TC Heaney Needle Holders, OR Grade, Sklar
Supplier: Sklar
21-8060 21-8061
89023-390EA 536.31 USD InStock
89023-390 89023-392
TC Heaney Needle Holders, OR Grade, Sklar
Forceps
Sklar® TC Heaney Needle Holders are used for light to medium weight suturing, most commonly in gynecological procedures such as vaginal and abdominal hysterectomy, and dilation and curettage.
The slightly curved shanks and tips provide the user a better view when suturing intravaginally, and the cross-serrations provide a secure hold on the needle
Censitrac Ready products are industry standard and constructed of stainless steel.
Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty