Would you like to log in again?
TC Bozeman-Heaney Needle Holder, OR Grade, Sklar®
Supplier: Sklar
21-8062
89023-394EA 602.77 USD InStock
89023-394
TC Bozeman-Heaney Needle Holder, OR Grade, Sklar®
Forceps
The Sklar® Tungsten Carbide Bozeman Needle Holder is commonly used for suturing in gynecological procedures
The shanks of this instrument are slightly curved and therefore provide greater visibility when used in the uterine cavity. Cross-serrated tips and a ratchet mechanism securely hold needles in place. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. This product features an "S" curve (slightly curved shanks with jaws slightly curved opposite direction) with cross-serrated tips and a length of 10-1/2 inches. TC instruments are identified by gold handles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty