Would you like to log in again?
Econo™ Webster Needle Holder, Floor Grade, Sklar
Supplier: Sklar
Econo™
21-809
89023-410PK 105.55 USD InStock
89023-410
Econo™ Webster Needle Holder, Floor Grade, Sklar
Forceps
Econo™ Webster Needle Holder is used with small needles and fine suture material, in plastic surgery or subcutaneous procedures.
They are available with smooth or serrated jaws. Smooth jaws will cause the least damage to the needle; however, serrated jaws hold the needle more tightly. Physician preference determines the type of working surface utilized.
Econo™ instruments are floor-grade, disposable instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty