Would you like to log in again?
Econo™ Halsey Needle Holder, Floor Grade, Sklar®
Supplier: Sklar
21-810
89023-412PK 108.36 USD InStock
89023-412
Econo™ Halsey Needle Holder, Floor Grade, Sklar®
Forceps
Econo™ Halsey Needle Holders are used for holding small needles for suturing, most commonly during plastic surgery and procedures with delicate tissue, such as dermatology.
Econo™ Halsey Needle Holders are available with smooth or delicately serrated jaws. Smooth jaws will cause the least damage to the needle, however serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty