Would you like to log in again?
Econo™ Derf Needle Holder, Floor Grade, Sklar
Supplier: Sklar
Econo™
21-826
89023-414PK 103.05 USD InStock
89023-414
Econo™ Derf Needle Holder, Floor Grade, Sklar
Forceps
Econo™ Derf Needle Holders feature short jaws and delicate serrations, that hold small needles and 5-0 and 6-0 sutures.
For use in ophthalmic, dental, or plastic procedures, the shorter length allows the Derf Needle Holder to be controlled in smaller surgical areas.
Econo™ instruments are floor-grade, disposable instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty