Would you like to log in again?
Econo™ Baumgartner Needle Holder, Floor Grade, Sklar
Supplier: Sklar
Econo™
21-832
89023-416PK 107.75 USD InStock
89023-416
Econo™ Baumgartner Needle Holder, Floor Grade, Sklar
Forceps
Econo™ Baumgartner Needle Holder features heavy serrations, which make this instrument capable of holding larger needles and dense suture material
Econo™ instruments are floor-grade, disposable instruments.
They are used in procedures on the trunk and limbs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty