Would you like to log in again?
Econo™ Crile-Wood Needle Holder, Floor Grade, Sklar
Supplier: Sklar
Econo™
21-838
89023-418PK 110.24 USD InStock
89023-418
Econo™ Crile-Wood Needle Holder, Floor Grade, Sklar
Forceps
Econo™ Crile-Wood Needle Holders are used to hold and guide small to medium size needles and suture materials.
The Crile-Wood Needle Holder has a gently tapered, blunt tip and is more delicate than the Mayo-Hegar but slightly less delicate than Webster, Halsey, or DeBakey needle holders. The ratcheted finger ring handle aids in securing needles for both continuous and interrupted closures.
Econo™ instruments are Sklar's floor-grade, disposable instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty