Would you like to log in again?
Econo™ Mayo-Hegar Needle Holder, Floor Grade, Sklar
Supplier: Sklar
Econo™
21-847 21-869 21-853 21-842
89023-424PK 160.94 USD InStock
89023-424 89023-426 89023-420 89023-422
Econo™ Mayo-Hegar Needle Holder, Floor Grade, Sklar
Forceps
The Econo™ Mayo-Hegar Needle Holder is a ratcheted, finger ring instrument available in a wide range of lengths, and used in multiple types of surgical procedures
Shorter length products may be used in oral, gynecological, or dermatological procedures, as well as with small animals. Larger lengths of this product may be used for deep tissue suturing such as in bariatric procedures, or in large animal procedures. The Mayo-Hegar needle holder is commonly used with larger, heavier suture needles because of it's wide jaws.
Econo™ instruments are disposable instruments.
Due to its versatility, it is one of the most commonly used surgical tools for both human and animal procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty