Would you like to log in again?
Surgi-OR™ TC Crile-Wood Needle Holder, Physician Grade, Sklar
Supplier: Sklar
95-137
89023-498EA 146.5 USD InStock
89023-498
Surgi-OR™ TC Crile-Wood Needle Holder, Physician Grade, Sklar
Forceps
The Surgi-OR™ TC Crile-Wood Needle Holder is used to hold and guide small to medium size needles and suture materials.
The Crile-Wood Needle Holder has a gently tapered, blunt tip and is generally more delicate than the Mayo-Hegar but slightly less delicate than Webster, Halsey, or DeBakey needle holders. The ratcheted finger ring handle aids in securing needles for both continuous and interrupted closures. Hand-crafted in Pakistan, Surgi-OR™ TC instruments are forged from the highest quality Japanese steel. They provide reliable instrumentation for the operating room and other surgical centers, and are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. Surgi-OR™ instruments are a cost-effective line of instruments that must meet Sklar's quality standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty