Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Surgi-OR™ Graves Vaginal Speculum, Mid Grade, Sklar
Supplier: Sklar
Surgi-OR™
95-312 95-324 95-322 95-303 95-304
89023-568EA 62.38 USD InStock
89023-568 89023-558 89023-564 89023-570 89023-560
Surgi-OR™ Graves Vaginal Speculum, Mid Grade, Sklar
Specula
Surgi-OR™ Graves Vaginal Speculum is a gynecological instrument used for performing pelvic examinations, pap smears, and examining the cervix.
  • Latex-Free
  • It is a double blade duckbill speculum inserted with the blades closed
The blades are then opened by an external lever and screw. The Graves Speculum has wider blades than the Pederson Speculum and the practitioner can best determine which type and size speculum is most appropriate for the patient.
Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from high quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty