Would you like to log in again?
Surgi-OR™ Halsted Mosquito Forceps, Mid-Grade, Sklar
Supplier: Sklar
Surgi-OR™
95-426
89023-614EA 39.44 USD InStock
89023-614
Surgi-OR™ Halsted Mosquito Forceps, Mid-Grade, Sklar
Forceps
Surgi-OR™ Halsted Mosquito Forceps are used in multiple procedures, as a hemostat for small blood vessels.
The ratcheted finger ring handle provides a secure, locking grip to halt blood flow. They are not appropriate for use with large vessels or arteries. A toothed variation is available for grasping delicate tissue in skin grafting, biopsies, or ophthalmologic procedures. Mosquito forceps are known for their fine tips, and short, fully serrated jaws. They are available curved or straight. The standard length for this instrument is 5", although Sklar does offer some with a length of 5 1/2".
Halsted Mosquito Forceps are very similar to Hartmann Mosquito Forceps, however, the Hartmann style is finer and more delicate. Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from high quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty