Would you like to log in again?
Surgi-OR™ Hartmann Mosquito Forceps, Mid Grade, Sklar
Supplier: Sklar
Surgi-OR™
95-431 95-421 95-409
89023-616EA 38.01 USD InStock
89023-616 89023-610 89023-612
Surgi-OR™ Hartmann Mosquito Forceps, Mid Grade, Sklar
Forceps
Surgi-OR™ Hartmann Mosquito Forceps are used as both a hemostat and for fine tissue dissection in shallow wounds and procedures
Known for their fine tips, short, and fully serrated jaws, these Mosquito Forceps can be used as a clamping instrument to halt blood flow so that cauterization or ligation can be performed. They are also used for holding small, fine sutures. Hartmann Mosquito Forceps are shorter and heavier than Halstead Mosquito Forceps.
Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from high quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers.
Mostly used in plastic and vascular surgeries, Hartmann Mosquito Forceps can also be used in pediatric and ophthalmologic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty