Would you like to log in again?
Surgi-OR™ Kelly Hemostatic Forceps, Mid Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Surgi-OR™
95-442 95-436
89023-620EA 36.86 USD InStock
89023-620 89023-618
Surgi-OR™ Kelly Hemostatic Forceps, Mid Grade, Sklar
Forceps
Surgi-OR™ Kelly Hemostatic Forceps are used for clamping large blood vessels or manipulating heavy tissue
They are available with a ratcheted finger ring handle. Similar to Rankin forceps, the jaws of Kelly Forceps are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws, however, Kelly forceps are shorter and heavier than Rankin forceps.
Surgi-OR™ products are mid-grade instruments suitable for the operating room and other surgical centers.
They may be used for soft tissue dissection.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty