Would you like to log in again?
Surgi-OR™ Rochester-Pean Hemostatic Forceps, Mid Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Surgi-OR™
95-473 95-485 95-471 95-477 95-494 95-470 95-492 95-483 95-481
89023-636EA 40.07 USD InStock
89023-636 89023-648 89023-638 89023-644 89023-634 89023-646 89023-650 89023-640 89023-642
Surgi-OR™ Rochester-Pean Hemostatic Forceps, Mid Grade, Sklar
Forceps
Surgi-OR™ Rochester-Pean Hemostatic forceps are used for clamping larger tissue and vessels for hemostasis.
The full horizontal serrations and availability in multiple lengths make these forceps a versatile instrument used in multiple procedures. They are available curved or straight.
Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from high quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty