Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Surgi-OR™ Rochester-Ochsner Forceps, Mid-Grade, Sklar
Supplier: Sklar
Surgi-OR™
95-525 95-517 95-501 95-514 95-518 95-512 95-500 95-515 95-521 95-516 95-504 95-513
89023-658EA 49.3 USD InStock
89023-658 89023-654 89023-666 89023-656 89023-668 89023-672 89023-662 89023-674 89023-652 89023-664 89023-670 89023-660
Surgi-OR™ Rochester-Ochsner Forceps, Mid-Grade, Sklar
Forceps
Surgi-OR™ Rochester-Oschner Forceps are a heavy instrument designed to grasp medium to heavy tissue or occlude heavy, dense vessels.
They have horizontal serrations the entire length of the jaw as well as 1 x 2 teeth at the tip. The combination of full serrations plus teeth ensure a firm grip on the tissue or vessel being held. Rochester-Oschner Forceps are available either curved or straight and in multiple lengths.
Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty