Would you like to log in again?
Surgi-OR™ Lorna (Edna) Towel Clamps, OR Grade, Sklar®
Supplier: Sklar
95-629 95-635
89023-688EA 45.91 USD InStock
89023-688 89023-690
Surgi-OR™ Lorna (Edna) Towel Clamps, OR Grade, Sklar®
Forceps
Surgi-OR™ Lorna (Edna) Towel Clamps are a non-perforating clamp used to secure instruments, surgical materials such as tubing, and towels to the drape surrounding a surgical site
The jaws and tips are curved while the shanks and handle are flat so they can rest on the patient. The teeth on the tips of this towel clamp are blunted so they grip tightly but do not penetrate the surgical drape. They also have ratcheted finger ring handles so a firm hold can be secured. The Lorna Towel Clamp is also commonly referred to as the Edna Towel Clamp. This clamp should never be used on skin. Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR™ instruments are a cost-effective line of instruments that must meet Sklar's quality standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty