Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Surgi-OR™ Adson Bayonet Dressing Forceps, Mid Grade, Sklar
Supplier: Sklar
Surgi-OR™
95-772 95-771
89023-710EA 30.57 USD InStock
89023-710 89023-712
Surgi-OR™ Adson Bayonet Dressing Forceps, Mid Grade, Sklar
Forceps
Surgi-OR™ Adson Bayonet Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds
They have a wide thumb grasp for increased precision and control. Surgi-OR™ mid-grade instruments provide cost-effective, reliable instrumentation for the operating room and surgical centers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty