Would you like to log in again?
Surgi-OR™ TC Derf Needle Holders, Mid Grade, Sklar®
Supplier: Sklar
Surgi-OR™
95-136 95-826
89023-496EA 153.5 USD InStock
89023-496 89023-728
Surgi-OR™ TC Derf Needle Holders, Mid Grade, Sklar®
Forceps
Surgi-OR™ TC Derf Needle Holders feature short jaws and delicate serrations that hold small needles and commonly 5-0 and 6-0 sutures
Hand-crafted in Pakistan, Surgi-OR™ TC mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. They are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.
For use in ophthalmic, dental, or plastic procedures, the shorter length allows the Derf Needle Holder to be controlled in smaller surgical areas.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty