Would you like to log in again?
Econo™ Sterile Iris Scissors, Floor Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Econo™
96-2651 96-2505 96-2504 96-2507 96-2506 96-2499 96-2497 96-2650
89023-768CS 328.93 USD InStock
89023-768 89023-758 89023-958 89023-764 89023-766 89023-960 89023-770 75995-370
Econo™ Sterile Iris Scissors, Floor Grade, Sklar
Scissors
Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments.
  • Ready-to-use
  • Eliminates the need for cleaning and reprocessing
  • Significantly reduce likelihood of cross-contamination
These fine, sharp scissors are designed for detailed dissection of fine tissue and fine suture removal
Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments made in Pakistan. Econo™ Sterile instruments eliminate the need for cleaning and reprocessing. Because they are both sterile and disposable, Econo™ Sterile instruments significantly reduce the likelihood of cross-contamination.
In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty