Would you like to log in again?
Econo™ Sterile Lister Bandage Scissors, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Econo™
96-2501 96-2503 96-2500 96-2502 96-2654
89023-966CS 774.43 USD InStock
89023-966 75995-372 75995-374 89023-760 89023-762
Econo™ Sterile Lister Bandage Scissors, Sklar
Scissors
Econo™ Sterile Lister Bandage Scissors are a versatile instrument with multiple uses.
  • Ready-to-use
  • Angled jaws
  • Lower blade is slightly longer
  • Flattened blunt nodule on tip of lower blade
The scissors are commonly used for sizing dressings and removing circumferential bandages. They may also be used to cut tougher material such as plaster, fabric and occasionally umbilical cord. The jaws of the Lister Bandage Scissors are angled with the lower blade being slightly longer. The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without harming the skin. This feature allows Lister Bandage Scissors to occasionally be used during cesarean section to open the uterus without causing inadvertent injury to the baby. Ring handles can be equal in size or with one ring larger. Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments sold in bulk quantities.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty