Would you like to log in again?
Econo™ Sterile Spencer (Littauer) Suture Scissors, Sklar
Supplier: Sklar
96-2511
89023-774CS 200.36 USD InStock
89023-774
Econo™ Sterile Spencer (Littauer) Suture Scissors, Sklar
Scissors
Econo™ Sterile Spencer (Littauer) Suture Scissors are used for suture removal
These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the scissor blade prior to cutting. Littauer suture scissors typically have larger, less delicate tips than Spencer Stitch Scissors, and are often used for removing larger sutures. They should not be used for cutting tissue. Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments sold in bulk quantities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty