Would you like to log in again?
Econo™ Sterile Hartmann Mosquito Forceps, Floor Grade, Sklar
Supplier: Sklar
Econo™
96-2556 96-2554 96-2557 96-2555
89023-856CS 371.91 USD InStock
89023-856 89023-858 89023-854 89023-860
Econo™ Sterile Hartmann Mosquito Forceps, Floor Grade, Sklar
Forceps
Econo™ Sterile Hartmann Mosquito Forceps are used as both a hemostat and for fine tissue dissection in shallow wounds and procedures
Thanks to their fine tips, short, and fully serrated jaws, these Mosquito Forceps can be used as a clamping instrument to halt blood flow so that cauterization or ligation can be performed. They are also used for holding small, fine sutures. Hartmann Mosquito Forceps are shorter and heavier than Halstead Mosquito Forceps.
Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments made in Pakistan. Econo™ Sterile instruments eliminate the need for cleaning and reprocessing. Because they are both sterile and disposable, Econo™ Sterile instruments significantly reduce the likelihood of cross-contamination.
Most commonly utilized in plastic and vascular surgeries, Hartmann Mosquito Forceps can also be used in pediatric and ophthalmologic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty