Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Econo™ Sterile Kelly Hemostatic Forceps, Floor Grade, Sklar
Supplier: Sklar
Econo™
96-2563 96-2560 96-2652 96-2653 96-2561 96-2562
89023-868CS 179.08 USD InStock
89023-868 89023-962 89023-964 89023-866 89023-870 89023-872
Econo™ Sterile Kelly Hemostatic Forceps, Floor Grade, Sklar
Forceps
Econo™ Sterile Kelly Hemostatic Forceps are used for clamping large blood vessels or manipulating heavy tissue. They may also be used for soft tissue dissection.
  • Ready-to-use
  • Available straight or curved
  • Ratcheted finger ring handle
The forceps are available curved or straight with a ratcheted finger ring handle
Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments.
The jaws of Kelly Forceps are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty