Would you like to log in again?
Econo™ Sterile Baumgartner Needle Holder, Floor Grade, Sklar
Supplier: Sklar
Econo™
96-2581 96-2580
89023-904CS 407.43 USD InStock
89023-904 89023-906
Econo™ Sterile Baumgartner Needle Holder, Floor Grade, Sklar
Forceps
Econo™ Sterile Baumgartner Needle Holder is used for holding suture needles.
It has shorter jaws than both Crile-Wood and Mayo-Hegar Needle Holders. Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments made in Pakistan.
Econo™ Sterile instruments eliminate the need for cleaning and reprocessing. Because they are both sterile and disposable, Econo™ Sterile instruments significantly reduce the likelihood of cross-contamination.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty