Would you like to log in again?
Econo™ Sterile Halsey Needle Holders, Floor Grade, Sklar
Supplier: Sklar
Econo™
96-2585 96-2584
89023-912CS 467.76 USD InStock
89023-912 89023-914
Econo™ Sterile Halsey Needle Holders, Floor Grade, Sklar
Forceps
Econo™ Sterile Halsey Needle Holders are used for holding small needles for suturing, during plastic surgery and procedures with delicate tissue, such as dermatology.
They are available with smooth or delicately serrated jaws. Smooth jaws will cause the least damage to the needle; however, serrated jaws hold the needle more tightly. Physician preference determines the type of working surface utilized.
Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty