Would you like to log in again?
Econo™ Spencer Stitch Scissors, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Econo™
96-2596 96-2663 96-2672 21-352 96-2598
89023-934CS 280.67 USD InStock
89023-934 89023-170 89023-984 75995-416 89023-938
Econo™ Spencer Stitch Scissors, Sklar
Scissors
Econo™ Spencer Stitch Scissors are primarily used for suture removal.
  • Small hook-shaped tip on one blade
  • Delicate tip for removing smaller sutures
These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. They typically have smaller, more delicate tips that are used for removing smaller sutures. These scissors should not be used for cutting tissue.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty