Would you like to log in again?
Econo™ Sterile Bozeman Forceps, Floor Grade, Sklar
Supplier: Sklar
Econo™
96-2622 96-2620
89023-940CS 1227.21 USD InStock
89023-940 89023-942
Econo™ Sterile Bozeman Forceps, Floor Grade, Sklar
Forceps
Econo™ Sterile Bozeman Forceps are a multi-functional instrument commonly used in gynecological procedures
The shanks of this instrument are slightly curved and therefore provide greater visibility when used in the uterine cavity. The jaws are available either curved or straight.
Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments made in Pakistan. Econo™ Sterile instruments eliminate the need for cleaning and reprocessing. Because they are both sterile and disposable, Econo™ Sterile instruments significantly reduce the likelihood of cross-contamination.
They are used for tissue removal in dilation and curettage procedures, intrauterine device retrieval, and packing the uterine cavity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty