Would you like to log in again?
Econo™ Sterile Babcock Forceps, Floor Grade, Sklar
Supplier: Sklar
Econo™
96-2705 96-2706
89024-014CS 695.8 USD InStock
89024-014 89024-012
Econo™ Sterile Babcock Forceps, Floor Grade, Sklar
Forceps
Econo™ Sterile Babcock Forceps are finger ring, ratcheted, non-perforating forceps used to grasp delicate tissue
Babcock Forceps are similar to Allis forceps; however, may be considered less traumatic due to their wider, rounded grasping surface. The jaws are circumferential, and the tips are triangular and fenestrated with horizontal serrations. They are particularly useful for grasping tube-shaped structures.
Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments.
They are used with intestinal and laparotomy procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty