Would you like to log in again?
Econo™ Sterile Metzenbaum-Lahey Dissecting Scissors, Sklar
Supplier: Sklar
96-2720 96-2517 96-2721 96-2516
89023-782CS 490.58 USD InStock
89023-782 89023-784 89024-024 89024-022
Econo™ Sterile Metzenbaum-Lahey Dissecting Scissors, Sklar
Scissors
Econo™ Sterile Metzenbaum Dissecting Scissors are most commonly used for dissecting and cutting tissue
These scissors are not recommended for cutting sutures, drains, or heavy tissue. Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor grade instruments made in Pakistan. Econo™ Sterile instruments eliminate the need for cleaning and reprocessing, as they are both sterile and disposable. These instruments significantly reduce the likelihood of cross-contamination. They are available in bulk quantities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty