Would you like to log in again?
Static Shielding Bags 1000, Desco Industries
Supplier: Desco Industries
3M™
10068 3002024 100616 1002020 100614 3001024 100912 3001010 300630 300510 100426 10048 1001026 1001430 300610 10026 1002224 3002424 100818 100626 30088 1001114 10024 30046 1001830 3001624 100716 100624 100618 100715 10023 3001818 300624 300715 1001212 3001418 30033 300430 10056 10069 1006.516 30048 3001030 1003024 100710 100424 300424 10046 1001214 1001225 1001018 100612 3001216 30035 100830 100711 100810 1001014
89024-192PK 39.32 USD InStock
89024-192 89024-072 89024-190 89024-070 89024-114 89024-236 89024-038 89024-118 89024-032 89024-194 89024-030 89024-198 89024-110 89024-078 89024-232 89024-082 89024-248 89024-204 89024-128 89024-246 89024-202 89024-048 89024-126 89024-208 89024-206 89024-240 89024-086 89024-042 89024-164 89024-040 89024-084 89024-044 89024-088 89024-242 89024-166 89024-122 89024-170 89024-092 89024-136 89024-218 89024-178 89024-058 89024-056 89024-210 89024-182 89024-028 89024-226 89024-106 89024-224 89024-104 89024-064 89024-062 89024-184 89024-068 89024-146 89024-066 89024-100 89024-188
Static Shielding Bags 1000, Desco Industries
Bags Storage Bags
These transparent metal-in static shielding bags are designed to provide a static safe environment for sensitive electronic devices.
  • Bags are printed with an ESD warning symbol and a lot code for traceability
  • Open top and ZipTop styles are available
  • Bags are heat sealable
These bags are manufactured from a polyester, metal, polyethylene laminate. The polyester dielectric in concert with the metal layer is intended to provide shielding of Electrostatic Discharge (ESD) and to help minimize the penetration of electric field.
Certifications: These bags are tested to meet or exceed certain electrical and physical requirements of Static Shielding Bags, ANSI/ESD S541, and to be ANSI/ESD S20.20 program compliant. Bags are RoHS Compliant and lead-free.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty