These transparent metal-in static shielding bags are designed to provide a static safe environment for sensitive electronic devices.
- Bags are printed with an ESD warning symbol and a lot code for traceability
- Open top and ZipTop styles are available
- Bags are heat sealable
These bags are manufactured from a polyester, metal, polyethylene laminate. The polyester dielectric in concert with the metal layer is intended to provide shielding of Electrostatic Discharge (ESD) and to help minimize the penetration of electric field.
Certifications: These bags are tested to meet or exceed certain electrical and physical requirements of Static Shielding Bags, ANSI/ESD S541, and to be ANSI/ESD S20.20 program compliant. Bags are RoHS Compliant and lead-free.