Would you like to log in again?
BAG STATIC MTALOUT 12X16 PK100
Supplier: DESCO
Bag^ SCC*1500 Metal-out static shielding bags. Regular (flat) opening. 12 inch x16 inch inside dimensions. Sold 100 bags per pack.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BAG STATIC MTALOUT 12X16 PK100 89024-278 1501216 Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty