Would you like to log in again?
Dri-Shield® Moisture Barrier Bag, 2000 Series, SCS, Desco Industries
Supplier: Desco Industries
70035
89024-310PK 27.08 USD InStock
89024-310
Dri-Shield® Moisture Barrier Bag, 2000 Series, SCS, Desco Industries
Bags Sample Bags
This aluminized moisture barrier bag is designed to provide packaging for ESD and moisture sensitive items, both inside and outside an ESD protected area.
  • Heat sealable and suitable for vacuum packaging
  • Printed with ESD and moisture warning symbols and a lot code for traceability
  • Manufactured from a laminate of multiple layers of aluminized polyester and polyethylene
Metal layers are intended to provide shielding of Electrostatic discharge (ESD) and to help minimize the penetration of electric field while providing moisture protection. Dissipative inner and outer surfaces allows bag to be grounded when placed on a grounded surface or handled by a grounded operator.
Certifications: Meets ANSI/ESD S20.20, ANSI/ESD S541, EIA 541, JESD625-A, and EIA 583.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty