Would you like to log in again?
BAG BARRIER 3.6MIL 6X8 PK 100
Supplier: DESCO
Bag^ SCC* Dri-Shield 2000 Moisture Barrier Bag, Designed for dry packaging of electronic devices. 3.6 mil. 6 inch x8 inch inside dimensions, Sold 100 bags per pack.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BAG BARRIER 3.6MIL 6X8 PK 100 89024-318 70068 Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty