Would you like to log in again?
Dri-Shield® Moisture Barrier Bags, 2000 Series, SCS, Desco Industries
Supplier: Desco Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7001518 7001020 7001216 7001824 7001012 700810
89024-326PK 55.69 USD InStock
89024-326 89024-338 89024-332 89024-344 89024-352 89024-330
Dri-Shield® Moisture Barrier Bags, 2000 Series, SCS, Desco Industries
Bags Storage Bags
This aluminized moisture barrier bag is designed to provide packaging for ESD and moisture sensitive items, both inside and outside an ESD protected area. The bags are heat sealable and suitable for vacuum packaging.
  • 0.0036" (3.6 mil) thick bag film laminate constructed from multiple layers of aluminized polyester and polyethylene
  • Designed for moisture and electrostatic discharge (ESD) sensitive items (such as SMT)
  • < 0.03 Grams Moisture Vapor Transmission Rate (MVTR); designed for dry packing of moisture sensitive devices and to protect contents from damage due to ESD events or EMI
  • <10 nJ Energy Penetration Tested per ANSI/ESD STM11.31; protects contents from ESD charges and discharges
  • >20lb Puncture Strength Average; provides physical protection for contents
  • Dissipative Inner and Outer Surfaces Rs 1×10⁴ to < 1×10¹¹ Ω per ANSI/ESD STM11.11; allows bag to be grounded when placed on a grounded surface or handled by a grounded operator
Polyester provides puncture resistance. Metal layers are intended to provide shielding of Electrostatic Discharge (ESD) and to help minimize the penetration of electric field while providing moisture protection.
Certifications: Lead-free RoHS 2, REACH and Conflict Minerals compliant and Made in the USA. Printed with ESD Protective Symbol and lot code information.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty