Would you like to log in again?
DESICCANT 16 TYVEK PK100
Supplier: DESCO
Dessicant^ SCC* Desiccant for dry packaging of electronic and other devices. Meets the requirements of EIA 583 and MIL-D-3464, Types I&II. Montmorillonte clay in tyvek pouch. 16 unit desiccant , 150 per container.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
DESICCANT 16 TYVEK PK100 89024-394 16PLDES150 Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty