Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Humidity Indicator, Desco Industries

Supplier: Desco Industries

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

51015HIC125 6HIC200 3HIC125 4HIC100
89024-422PK 25.35 USD InStock
89024-422 89024-418 89024-416 89024-420
Humidity Indicator, Desco Industries
Test Paper Indicator Paper
Humidity Indicator Cards (HICs) are printed with moisture sensitive spots which respond to various levels of humidity with a visible color change from blue to pink.

  • Accuracy based on ambient temperature of 75 °F
  • Tolerance is approximately 2.5% for each 10 °F away from 75 °F

The moisture sensitive spots are made of blotting paper impregnated with cobaltous chloride. The humidity inside dry packed barrier bags can be monitored by including an HIC inside. When the bag is opened, the card can be examined for proper dryness inside the bag.

4HIC100 is designed for dry packaging SMD's, color reference bar assists with accurate readings. 6HIC200 is used in a variety of dry packaging application, this card monitors humidity levels across the widest range. 3HIC125 is popular for military applications.

Certifications: 4HIC100 and 6HIC200 complies with EIA 581, MIL-P-116, and MIL-I-8835. 51060HIC125 meets IPC/JEDEC J-STD-033B for observability and accuracy. 3HIC125 complies with MS 20003-2, and MIL-P-116.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty