Would you like to log in again?
LABEL ESD JEDEC 4X4 100/RL
Supplier: DESCO
SCC* 113 LABEL^ Designed for dry packaging, Displays the warning symbol and message in strict accordance with IPC/JEDEC J-STD-033 and JEDEC113. These destructive type labels are tamper evident, letting you easily monitor whether or not a bag has been
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LABEL ESD JEDEC 4X4 100/RL 89024-430 113LABEL Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty