Would you like to log in again?
Attention Labels, Desco Industries
Supplier: Desco Industries
129LABEL
89024-432EA 11.82 USD InStock
89024-432
Attention Labels, Desco Industries
Labels
This 2" by 2" self-adhesive label is easy to use and provides the Mil-STD-129, RS-471 ESD sensitivity symbol and attention message to warn that ESD control precautions are needed when opening package and handling contents.
  • Clearly identify products, packaging and areas requiring static protective measures
  • Self-adhesive labels suitable for automatic dispensers and reel stock applications
  • Label rolls include backing and paper core
  • Permanent acrylic polymer-based adhesive for one-time use
  • Made in the United States of America
This permanent adhesive destructible type label is 'tamper evident', letting users easily monitor whether or not a package has been opened. Use label on unit packs, intermediate, and exterior containers (bags, boxes and totes) to ensure proper ESD warning. The ESD warning symbol and message in accordance with the military standard. A total of 500 labels per roll. Roll core is 3" in diameter.
Certifications: In accordance with MIL-STD-129N.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty