Would you like to log in again?
WRIST STRAP W/12FEET CORD
Supplier: DESCO
SCC* AMWS121M Wrist Strap^ With 12' cord is made from jewelry-quality stainless steel. The Flex-Metal style band uses adjustable links to assure a secure and comfortable fit. Industry standard 4mm snap. Cord lasts over 1 million flexes.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
WRIST STRAP W/12FEET CORD 89024-458 AMWS121M Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty