Would you like to log in again?
Economy Heel Grounder Assembly, SCS, Desco Industries
Supplier: Desco Industries
HGC1M-ECG
89024-528EA 13.96 USD InStock
89024-528
Economy Heel Grounder Assembly, SCS, Desco Industries
Grounders Heel and Toe Grounders
Heel Grounder Assembly features a two-layer sole and a 24" conductive ribbon.
  • Durable
  • Cost-effective compliance for high-volume grounder use
Green-colored hook and loop fastener provides visual assurance that Heel Grounders are being used. The elastic strap offers adjustability for most any size shoe.
Certifications: ESD-SP9.2, ESD TR53.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty