Would you like to log in again?
Low-Profile Common Ground Cord, SCS, Desco Industries
Supplier: Desco Industries
LPCGC151M
89024-542EA 19.14 USD InStock
89024-542
Low-Profile Common Ground Cord, SCS, Desco Industries
Grounders Grounding Wires
With a low-profile top design it reduces snagging of clothes and work instruments, this common point ground cord connects two wrist straps and a work surface to ground, helping ensure that all parts are at the same ground potential.
  • 15 foot ground cord with 10 mm stud snap
  • Two banana jacks provide two grounding points for wrist straps or carts
  • 10 ring terminal at end of ground cord allows easy attachment to electrical ground
  • Includes 1 megohm resistor
  • Made in the United States of America
Easy installation requires simply snapping cord to a mat or work surface. Plug in the wrist strap coil cord. Equipped with holes for permanent mounting to a bench.
Certifications: Meets ANSI/ESD S20.20 and Grounding standard ANSI/ESD S6.1.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty